Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Leucine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-(-)-Leucine H-D-Leu-OH

A14842-06 A14842-06 A14842-22 A14842-22 A14842-14
AAA14842-14EA 220.99 USD InStock
AAA14842-14 AAA14842-06 AAAA14842-22 CAAAAA14842-22 CAAAA14842-06
D-(-)-Leucine 99%
D(-)-Leucine


Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty