Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4-methylthiazole 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Methylthiazol-2-ylamine

A14847-14 A14847-06 A14847-06 A14847-22 A14847-22 A14847-14
AAA14847-14EA 54.49 USD InStock
AAA14847-14 AAA14847-06 CAAAA14847-22 CAAAA14847-14 AAA14847-22 CAAAA14847-06
2-Amino-4-methylthiazole 98+%
2-Amino-4-methylthiazole


Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 231…233 °C
Melting Pt: 42…46 °C
Density: 1,202 g/cm³ (50 °C)
Flash Pt: 107 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005329
CAS Number: 1603-91-4
EINECS: 216-505-6
Merck Index: 13,00449
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty