Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl valerate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethyl Pentanoate Valeric acid ethyl ester

A14854-0F A14854-0F A14854-AP A14854-AP
AAA14854-0FEA 299.99 USD InStock
CAAAA14854-AP AAA14854-0F AAA14854-AP CAAAA14854-0F
Ethyl valerate 98%
Ethyl valerate


Formula: C₇H₁₄O₂
Boiling Pt: 142…145 °C
Melting Pt: –92 to –90 °C
Density: 0.874
Flash Pt: 38 °C(100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS Number: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: "3",III
Merck Index: 12,10042
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty