Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloropyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

A14866-22 A14866-36 A14866-36 A14866-22
AAA14866-36EA 124.99 USD InStock
CAAAA14866-36 CAAAA14866-22 AAA14866-36 AAA14866-22
2-Chloropyridine 99%
2-Chloropyridine


Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 167…170 °C
Melting Pt: –46 °C
Density: 1.208
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006228
CAS Number: 109-09-1
EINECS: 203-646-3
UN: 2822
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty