Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Phenylpropionic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Hydrocinnamic acid

A14908-36 A14908-36 A14908-22 A14908-22
AAA14908-36EA 271.99 USD InStock
AAA14908-36 AAA14908-22 CAAAA14908-36 CAAAA14908-22
3-Phenylpropionic acid 99%
3-Phenylpropionic acid


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 280 °C
Melting Pt: 47…50 °C
Density: 1.071
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
EINECS: 207-924-5
Merck Index: 13,04805
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty