Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium tetraoxalate dihydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Oxalic acid, potassium salt dihydratePotassium trihydrogen dioxalate dihydrate

A14933-0B A14933-30 A14933-0I A14933-0B A14933-30 A14933-0I
AAA14933-0BEA 134.99 USD InStock
CAAAA14933-30 AAA14933-0B CAAAA14933-0I CAAAA14933-0B AAA14933-30 AAA14933-0I
Potassium tetraoxalate dihydrate 98+%
Potassium tetraoxalate dihydrate
Applications: Metal polishing determination of pH


Formula: C₄H₃KO₈·2H₂O
MW: 254.19 (218.16 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150443
CAS Number: 6100-20-5
EINECS: 204-874-6
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty