Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl propionate 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propionic acid methyl ester Propanoic acid methyl ester

A14944-AP A14944-AP A14944-AE A14944-0F A14944-AE A14944-0F
AAA14944-0FEA 410.99 USD InStock
AAA14944-0F AAA14944-AE CAAAA14944-0F AAA14944-AP CAAAA14944-AP CAAAA14944-AE
Methyl propionate 99%
Methyl propionate


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 78…80 °C
Melting Pt: –88 °C
Density: 0.915
Flash Pt: -2 °C(28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009306
CAS Number: 554-12-1
EINECS: 209-060-4
UN: 1248
ADR: "3",II
Merck Index: 12,06191
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty