Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Camphorquinone 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dioneDL-Camphoroquinone

A14967-22 A14967-06 A14967-22 A14967-06 A14967-14 A14967-14
AAA14967-14EA 251.99 USD InStock
AAA14967-14 AAA14967-06 CAAAA14967-22 AAA14967-22 CAAAA14967-14 CAAAA14967-06
(±)-Camphorquinone 99%
(±)-Camphorquinone


Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00064160
CAS Number: 10373-78-1
EINECS: 233-814-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty