Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichlobenil 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,6-Dichlorobenzonitrile

A14986-22 A14986-22
AAA14986-22EA 149.99 USD InStock
CAAAA14986-22 AAA14986-22
Dichlobenil 98%
Dichlobenil


Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Boiling Pt: 270 °C
Melting Pt: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00001781
CAS Number: 1194-65-6
EINECS: 214-787-5
UN: 2811
ADR: "6.1",III
Merck Index: 13,03068
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty