Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocyclopropane 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cyclopropyl bromide

A14996-30 A14996-09 A14996-09 A14996-30 A14996-18 A14996-18
AAA14996-09EA 71.19 USD InStock
AAA14996-09 CAAAAA14996-30 AAAA14996-30 AAA14996-18 CAAAA14996-09 CAAAA14996-18
Bromocyclopropane 99%
Bromocyclopropane


Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 68…70 °C
Density: 1.515
Flash Pt: -6 °C(21 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001271
CAS Number: 4333-56-6
EINECS: 224-375-7
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty