Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloroquinaldine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Chloro-2-methylquinoline

A15002-06 A15002-14 A15002-06 A15002-14 A15002-03 A15002-03
AAA15002-06EA 82.58 USD InStock
CAAAA15002-14 CAAAA15002-03 AAA15002-06 AAA15002-14 AAA15002-03 CAAAA15002-06
4-Chloroquinaldine 97%
4-Chloroquinaldine


Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Boiling Pt: 269…270 °C
Melting Pt: ca 25 °C
Density: 0,881
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006757
CAS Number: 4295-06-1
EINECS: 224-300-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty