Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2,6-dichloropyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,6-Dichloro-4-pyridylamine 2,6-Dichloropyridin-4-amine

A15030-06 A15030-03 A15030-06 A15030-MD A15030-03
AAA15030-03EA 41.08 USD InStock
AAA15030-03 CAAAA15030-06 AAA15030-06 CAAAA15030-03 CAAAAA15030-MD
4-Amino-2,6-dichloropyridine 98+%
4-Amino-2,6-dichloropyridine


Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00052832
CAS Number: 2587-02-2
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty