Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Malononitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

A15046-0E A15046-0E A15046-36 A15046-22 A15046-36 A15046-22
AAA15046-22EA 30.12 USD InStock
CAAAA15046-0E AAA15046-22 CAAAA15046-36 AAA15046-36 AAA15046-0E CAAAA15046-22
Malononitrile 99%
Malononitrile
Crosslinking agent for modification of proteins via amidation


Formula: C₃H₂N₂
MW: 66.06 g/mol
Boiling Pt: 220…222 °C
Melting Pt: 30…34 °C
Density: 1.049
Flash Pt: 112 °C(233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: "6.1",II
Merck Index: 13,05733
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty