Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxyphenyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

A15090-22 A15090-14 A15090-22 A15090-06 A15090-14 A15090-06
AAA15090-14EA 162.29 USD InStock
CAAAA15090-06 CAAAA15090-14 AAA15090-14 AAA15090-22 AAA15090-06 CAAAA15090-22
3-Methoxyphenyl isothiocyanate 98%
3-Methoxyphenyl isothiocyanate


Formula: C₈H₇NOS
MW: 165.22 g/mol
Boiling Pt: 133…134 °C/10 mm
Density: 1.179
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040335
CAS Number: 3125-64-2
EINECS: 000-000-0
UN: 2922
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty