Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxyphthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A15091-22 A15091-06 A15091-14 A15091-06 A15091-14 A15091-22
AAA15091-14EA 500.99 USD InStock
AAA15091-14 CAAAA15091-06 AAA15091-22 CAAAA15091-14 AAA15091-06 CAAAA15091-22
4-Hydroxyphthalic acid 98%
4-Hydroxyphthalic acid


Formula: C₈H₆O₅
MW: 182.13 g/mol
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: ca 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013984
CAS Number: 610-35-5
EINECS: 210-221-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty