Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Tolylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-Methylphenyl)acetic acid

A15129-22 A15129-22 A15129-06 A15129-06 A15129-14 A15129-14
AAAA15129-14EA 75.99 USD InStock
AAAA15129-14 AAA15129-06 CAAAA15129-22 CAAAA15129-14 AAA15129-22 CAAAA15129-06
p-Tolylacetic acid 99%
p-Tolylacetic acid


Formula: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 265…267 °C
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004353
CAS Number: 622-47-9
EINECS: 210-738-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty