Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Iodovanillin 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

A15162-18 A15162-09 A15162-30 A15162-18 A15162-30 A15162-09
AAA15162-18EA 176.99 USD InStock
CAAAA15162-18 AAA15162-18 AAA15162-09 CAAAA15162-30 AAA15162-30 CAAAA15162-09
5-Iodovanillin 98%
5-Iodovanillin


Formula: C₈H₇IO₃
MW: 278.05 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006941
CAS Number: 5438-36-8
EINECS: 226-617-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty