Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Diphenylguanidine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N,N'-Diphenylguanidine sym-Diphenylguanidine

A15234-22 A15234-0B A15234-22 A15234-30 A15234-30 A15234-0B
AAAA15234-22EA 26.59 USD InStock
AAAA15234-22 CAAAAA15234-22 CAAAA15234-30 AAA15234-0B AAA15234-30 CAAAA15234-0B
1,3-Diphenylguanidine 97%
1,3-Diphenylguanidine


Formula: C₁₃H₁₃N₃
MW: 211.27 g/mol
Boiling Pt: >170 °C (dec.)
Melting Pt: 146…150 °C
Density: 1.130
Flash Pt: 170 °C(338 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001758
CAS Number: 102-06-7
EINECS: 203-002-1
UN: 3077
ADR: "9",III
Merck Index: 13,03356
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty