Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Acetylbenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15239-03 A15239-14 A15239-06 A15239-03 A15239-06 A15239-14
AAA15239-06EA 151.99 USD InStock
CAAAA15239-03 CAAAA15239-14 AAA15239-06 AAA15239-14 AAA15239-03 CAAAA15239-06
4-Acetylbenzoic acid 98+%
4-Acetylbenzoic acid


Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00002561
CAS Number: 586-89-0
EINECS: 209-588-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty