Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-1H-1,2,4-triazole-5-thiol 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Amino-5-mercapto-1,2,4-triazole

A15242-22 A15242-14 A15242-06 A15242-14 A15242-22 A15242-06
AAA15242-22EA 401.99 USD InStock
CAAAA15242-14 AAA15242-22 CAAAA15242-22 AAA15242-14 AAA15242-06 CAAAA15242-06
3-Amino-1H-1,2,4-triazole-5-thiol 98+%
3-Amino-1H-1,2,4-triazole-5-thiol


Formula: C₂H₄N₄S
MW: 116.15 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005231
CAS Number: 16691-43-3
EINECS: 240-735-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty