Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methyl-1,2,3,4-tetrahydroquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,2,3,4-Tetrahydro-6-methylquinoline

A15245-18 A15245-18 A15245-09 A15245-09
AAA15245-18EA 744.99 USD InStock
CAAAA15245-18 CAAAA15245-09 AAA15245-18 AAA15245-09
6-Methyl-1,2,3,4-tetrahydroquinoline 98%
6-Methyl-1,2,3,4-tetrahydroquinoline


Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 262…263 °C/712 mm
Melting Pt: 33…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00023887
CAS Number: 91-61-2
EINECS: 202-083-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty