Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Indol-3-yl)butyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Indole-3-butyric acid

A15260-14 A15260-06 A15260-22 A15260-14 A15260-06 A15260-22
AAA15260-06EA 48.3 USD InStock
AAA15260-06 AAA15260-14 CAAAA15260-14 AAA15260-22 CAAAA15260-22 CAAAA15260-06
4-(Indol-3-yl)butyric acid 98%
4-(Indol-3-yl)butyric acid


Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Melting Pt: 121…125 °C
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: "6.1",III
Merck Index: 13,04987
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty