Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Formylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Carboxybenzaldehyde Terephthalaldehydic acid

A15277-18 A15277-18 A15277-30 A15277-30 A15277-09 A15277-09
AAA15277-30EA 686.99 USD InStock
AAA15277-30 AAA15277-09 CAAAA15277-09 AAA15277-18 CAAAA15277-18 CAAAA15277-30
4-Formylbenzoic acid 98%
4-Formylbenzoic acid


Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: ca 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00006951
CAS Number: 619-66-9
EINECS: 210-607-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty