Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(4-Iodophenyl)pyrrole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-(4-Iodophenyl)pyrrole

A15305-14 A15305-06 A15305-03 A15305-03 A15305-06
AAA15305-03EA 58.69 USD InStock
AAA15305-03 AAAA15305-14 CAAAA15305-06 CAAAA15305-03 AAA15305-06
1-(4-Iodophenyl)pyrrole 97%
1-(4-Iodophenyl)pyrrole


Formula: C₁₀H₈IN
MW: 269.08 g/mol
Melting Pt: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00052399
CAS Number: 92636-36-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty