Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,N-Trimethylanilinium tribromide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Phenyltrimethylammonium tribromide Trimethylphenylammonium tribromide

A15326-18 A15326-0B A15326-18 A15326-30 A15326-30 A15326-0B
AAA15326-30EA 296.99 USD InStock
AAA15326-30 CAAAA15326-18 AAAA15326-0B CAAAA15326-30 CAAAAA15326-0B AAA15326-18
N,N,N-Trimethylanilinium tribromide 97%
N,N,N-Trimethylanilinium tribromide


Formula: C₉H₁₄Br₃N
MW: 375.93 g/mol
Melting Pt: 112…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00011789
CAS Number: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty