Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methylnaphthalene 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: α-Methylnaphthalene

A15327-22 A15327-36 A15327-22 A15327-36
AAA15327-36EA 95.19 USD InStock
AAA15327-36 AAA15327-22 CAAAA15327-36 CAAAA15327-22
1-Methylnaphthalene 96%
1-Methylnaphthalene


Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 240…243 °C
Melting Pt: –22 °C
Density: 1.024
Flash Pt: 82 °C(179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty