Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Pyrrolidone 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pyrrolidin-2-one

A15332-36 A15332-36 A15332-A3 A15332-A3
AAAA15332-36EA 54.04 USD InStock
AAAA15332-36 AAA15332-A3 CAAAA15332-A3 CAAAA15332-36
2-Pyrrolidone 99%
2-Pyrrolidone


Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 245 °C
Melting Pt: 23…25 °C
Density: 1,12
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
EINECS: 210-483-1
Merck Index: 12,08200
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty