Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl oxalate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Oxalic acid dimethyl ester

A15359-22 A15359-36 A15359-22 A15359-0E A15359-36 A15359-0E
AAA15359-0EEA 570.99 USD InStock
CAAAA15359-22 AAA15359-0E AAA15359-36 AAA15359-22 CAAAA15359-36 CAAAA15359-0E
Dimethyl oxalate 99%
Dimethyl oxalate


Formula: C₄H₆O₄
MW: 118.09 g/mol
Boiling Pt: 162…163 °C
Melting Pt: 50…54 °C
Density: 1.148
Flash Pt: 75 °C(167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008442
CAS Number: 553-90-2
EINECS: 209-053-6
ADR: 8,III
Merck Index: 12,06181
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty