Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,6-Diiodohexane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Stabilization: Stabilised with copper

A15368-14 A15368-22 A15368-22 A15368-14
AAA15368-14EA 75.06 USD InStock
CAAAA15368-22 AAA15368-14 AAA15368-22 CAAAA15368-14
1,6-Diiodohexane 98% stabilized
1,6-Diiodohexane


Formula: C₆H₁₂I₂
MW: 337.97 g/mol
Boiling Pt: 123…128 °C/4 mm
Melting Pt: 10 °C
Density: 2.052
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001103
CAS Number: 629-09-4
EINECS: 211-073-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty