Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorothanesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (2-Chloroethyl)sulphonyl chloride

A15377-09 A15377-22 A15377-18 A15377-30 A15377-09 A15377-18 A15377-30 A15377-22
AAAA15377-22EA 381.99 USD InStock
CAAAA15377-18 AAAA15377-22 CAAAAA15377-22 CAAAA15377-09 AAA15377-18 CAAAA15377-30 AAA15377-09 AAA15377-30
2-Chlorothanesulphonyl chloride 98%
2-Chlorothanesulphonyl chloride


Formula: C₂H₄Cl₂O₂S
MW: 163.02 g/mol
Boiling Pt: 203 °C
Melting Pt: 3 °C
Density: 1.560
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007461
CAS Number: 1622-32-8
EINECS: 216-594-1
UN: 2927
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty