Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-4-nitrobenzene 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Chloronitrobenzene

A15396-30 A15396-0B A15396-0I
AAA15396-30EA 28.29 USD InStock
AAA15396-30 AAA15396-0I AAA15396-0B
1-Chloro-4-nitrobenzene 98+%
1-Chloro-4-nitrobenzene


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 238…242 °C
Melting Pt: 82…86 °C
Density: 1.515
Flash Pt: 127 °C(260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007285
CAS Number: 100-00-5
EINECS: 202-809-6
UN: 1578
ADR: "6.1",II
Merck Index: 13,02170
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty