Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Chloro-2-nitrobenzene2-Chloronitrobenzene

A15403-0B A15403-30 A15403-0I A15403-30 A15403-0I A15403-0B
AAA15403-0IEA 307.99 USD InStock
CAAAA15403-30 CAAAA15403-0B AAA15403-0I AAA15403-30 AAA15403-0B CAAAA15403-0I
1-Chloro-2-nitrobenzene 99%
1-Chloro-2-nitrobenzene


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 245…246 °C
Melting Pt: 31…34 °C
Density: 1.348
Flash Pt: 126 °C(258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007061
CAS Number: 88-73-3
EINECS: 201-854-9
UN: 1578
ADR: "6.1",II
Merck Index: 13,02170
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty