Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylvaleric acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Methylpentanoic acid Isocaproic acid

A15405-06 A15405-22 A15405-14 A15405-14 A15405-22 A15405-06
AAA15405-14EA 98.9 USD InStock
CAAAA15405-22 CAAAA15405-14 AAA15405-14 AAA15405-06 CAAAA15405-06 AAA15405-22
4-Methylvaleric acid 99%
4-Methylvaleric acid


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 199…201 °C
Melting Pt: –35 °C
Density: 0.923
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0
UN: 2922
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty