Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Heptylphenol 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-n-Heptylphenol

A15407-06 A15407-06 A15407-14 A15407-14 A15407-03 A15407-03
AAA15407-06EA 159.99 USD InStock
CAAAA15407-06 CAAAA15407-14 CAAAA15407-03 AAA15407-06 AAA15407-14 AAA15407-03
4-Heptylphenol 98+%
4-Heptylphenol


Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Boiling Pt: 156 °C/9 mm
Melting Pt: 25…28 °C
Density: 0.939 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041751
CAS Number: 1987-50-4
EINECS: 217-862-0
UN: 2430
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty