Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylthiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

A15428-22 A15428-06 A15428-14 A15428-22 A15428-14 A15428-06
AAA15428-14EA 68.99 USD InStock
CAAAA15428-06 CAAAAA15428-22 AAA15428-14 CAAAA15428-14 AAAA15428-22 AAA15428-06
4-Methylthiazole 99%
4-Methylthiazole


Formula: C₄H₅NS
Boiling Pt: 133…134 °C
Density: 1.093
Flash Pt: 32 °C(89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005340
CAS Number: 693-95-8
EINECS: 211-764-1
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty