Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Cyanobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A15434-06 A15434-22 A15434-06 A15434-22 A15434-14 A15434-14
AAA15434-22EA 433.99 USD InStock
CAAAA15434-14 AAA15434-22 CAAAA15434-06 CAAAA15434-22 AAA15434-06 AAA15434-14
4-Cyanobenzoic acid 98%
4-Cyanobenzoic acid


Formula: C₈H₅NO₂
MW: 147.13 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: ca 221 °C (dec.)
Density: 1,382 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002528
CAS Number: 619-65-8
EINECS: 210-606-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty