Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-p-toluidine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Amino-2-chlorotoluene 3-Chloro-4-methylaniline

A15439-0I A15439-30 A15439-0I A15439-0B A15439-30 A15439-0B
AAA15439-0IEA 371.33 USD InStock
CAAAA15439-30 AAA15439-0I CAAAA15439-0I AAA15439-30 CAAAA15439-0B AAA15439-0B
3-Chloro-p-toluidine 98%
3-Chloro-p-toluidine


Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 236…238 °C
Melting Pt: 22…24 °C
Density: 1.180
Flash Pt: 130 °C(266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007773
CAS Number: 95-74-9
EINECS: 202-446-3
UN: 2239
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty