Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethoxy)benzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(Trifluoromethoxy)benzene-1-sulphonyl chloride

A15467-03 A15467-06 A15467-03 A15467-14 A15467-06 A15467-14
AAA15467-03EA 36.89 USD InStock
AAA15467-03 AAA15467-14 CAAAA15467-06 AAA15467-06 CAAAA15467-03 CAAAA15467-14
4-(Trifluoromethoxy)benzenesulfonyl chloride 98%
4-(Trifluoromethoxy)benzenesulphonyl chloride


Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 120…122 °C/15 mm
Density: 1.536
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042408
CAS Number: 94108-56-2
EINECS: 302-397-9
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty