Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxybenzyl alcohol 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3-Methoxyphenyl)methanol 3-(Hydroxymethyl)anisole

A15471-14 A15471-14 A15471-22 A15471-22
AAA15471-22EA 166.99 USD InStock
AAA15471-22 AAA15471-14 CAAAA15471-14 CAAAA15471-22
3-Methoxybenzyl alcohol 98+%
3-Methoxybenzyl alcohol


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 252…254 °C
Melting Pt: 29…30 °C
Density: 1.112
Flash Pt: 109 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004637
CAS Number: 6971-51-3
EINECS: 230-200-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty