Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-fluorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A15478-06 A15478-06 A15478-14 A15478-14
AAA15478-14EA 249.99 USD InStock
AAA15478-14 AAA15478-06 CAAAA15478-06 CAAAA15478-14
2-Chloro-4-fluorobenzonitrile 98%
2-Chloro-4-fluorobenzonitrile


Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00042523
CAS Number: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty