Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-fluoroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

A15492-14 A15492-06 A15492-22 A15492-06 A15492-22 A15492-14
AAA15492-22EA 430.99 USD InStock
CAAAA15492-22 AAA15492-22 AAA15492-14 AAA15492-06 CAAAA15492-06 CAAAA15492-14
2-Chloro-4-fluoroaniline 97%
2-Chloro-4-fluoroaniline


Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 191…192 °C
Density: 1.219
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042530
CAS Number: 2106-02-7
EINECS: 218-282-0
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty