Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-1,3-difluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-2,4-difluorobenzene

A15495-18 A15495-30 A15495-09 A15495-18 A15495-30 A15495-09
AAA15495-09EA 24.99 USD InStock
CAAAA15495-18 CAAAA15495-09 AAA15495-09 CAAAA15495-30 AAA15495-18 AAA15495-30
4-Chloro-1,3-difluorobenzene 98%
4-Chloro-1,3-difluorobenzene


Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 126…127 °C
Melting Pt: –26 °C
Density: 1.353
Flash Pt: 32 °C(89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042569
CAS Number: 1435-44-5
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty