Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aminoacetaldehyde dimethyl acetal 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,2-Dimethoxyethanamine

A15498-22 A15498-36 A15498-22 A15498-36 A15498-14 A15498-14
AAA15498-36EA 521.99 USD InStock
AAA15498-36 AAA15498-14 CAAAA15498-22 AAA15498-22 CAAAA15498-36 CAAAA15498-14
Aminoacetaldehyde dimethyl acetal 99%
Aminoacetaldehyde dimethyl acetal


Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 135…138 °C
Density: 0.973
Flash Pt: 41 °C(106 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008135
CAS Number: 22483-09-6
EINECS: 245-026-5
UN: 2733
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty