Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenetole 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethoxybenzene Ethyl phenyl ether Phenoxyethane

A15501-AE A15501-0F A15501-AE A15501-0F A15501-AP A15501-AP
AAA15501-0FEA 268.99 USD InStock
CAAAA15501-AP AAA15501-0F AAA15501-AE CAAAA15501-0F AAA15501-AP CAAAA15501-AE
Phenetole 98+%
Phenetole


Formula: C₆H₅OC₂H₅
MW: 122.17 g/mol
Boiling Pt: 169…170 °C
Melting Pt: –30 °C
Density: 0.967
Flash Pt: 57 °C(134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009090
CAS Number: 103-73-1
EINECS: 203-139-7
UN: 3271
ADR: "3",III
Merck Index: 12,07374
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty