Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dibromo-5,5-dimethylhydantoin 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,3-Dibromo-5,5-dimethylimidazolidine-2,4-dione

A15510-0E A15510-0E A15510-36 A15510-36 A15510-22 A15510-22
AAA15510-36EA 61.37 USD InStock
AAA15510-36 AAA15510-0E CAAAA15510-36 CAAAA15510-22 AAA15510-22 CAAAA15510-0E
1,3-Dibromo-5,5-dimethylhydantoin 98%
1,3-Dibromo-5,5-dimethylhydantoin


Formula: C₅H₆Br₂N₂O₂
MW: 285.92 g/mol
Melting Pt: ca 198 °C (dec.)
Flash Pt: 155 °C(311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003189
CAS Number: 77-48-5
EINECS: 201-030-9
UN: 3085
ADR: "5.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty