Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-3-Chloroacrylic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A15527-MD A15527-03 A15527-06 A15527-06 A15527-03 A15527-MD
AAA15527-06EA 410.99 USD InStock
CAAAA15527-03 AAA15527-06 CAAAA15527-06 AAA15527-03 CAAAAA15527-MD AAAA15527-MD
trans-3-Chloroacrylic acid 99%
trans-3-Chloroacrylic acid


Formula: C₃H₃ClO₂
MW: 106.51
Melting Pt: 81…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064237
CAS Number: 2345-61-1
EINECS: 219-070-0
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty