Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Glucosamine hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride

A15532-18 A15532-30 A15532-18 A15532-0B A15532-0B A15532-30
AAA15532-0BEA 377.99 USD InStock
CAAAA15532-18 AAA15532-0B CAAAA15532-0B AAA15532-18 AAA15532-30 CAAAA15532-30
D-(+)-Glucosamine hydrochloride ≥98%
D-(+)-Glucosamine hydrochloride


Formula: C₆H₁₃NO₅
MW: 215.63 g/mol
Melting Pt: ca 192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00067674
CAS Number: 66-84-2
EINECS: 200-638-1
Order Now
Specification Test Results
Optical rotation +73° (c=1 in water, 20h)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty