Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriethylsilane 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triethylchlorosilane

A15547-09 A15547-18 A15547-30 A15547-09 A15547-30 A15547-18
AAA15547-09EA 69.82 USD InStock
AAA15547-09 CAAAA15547-30 AAA15547-18 AAA15547-30 CAAAA15547-18 CAAAA15547-09
Chlorotriethylsilane 98+%
Chlorotriethylsilane


Formula: C₆H₁₅ClSi
Boiling Pt: 145…147 °C
Melting Pt: ca …75 °C
Density: 0.899
Flash Pt: 29 °C(84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty