Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-4-nitroaniline 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-(Methylamino)-4-nitrobenzene

A15548-06 A15548-22 A15548-14
AAA15548-22EA 461.99 USD InStock
AAA15548-22 AAA15548-06 AAA15548-14
N-Methyl-4-nitroaniline 97%
N-Methyl-4-nitroaniline


Formula: C₇H₈N₂O₂
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 147…154 °C
Density: 1.201 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007305
CAS Number: 100-15-2
EINECS: 202-823-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty