Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitro-4,5-xylidine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4,5-Dimethyl-2-nitroaniline

A15556-14 A15556-22 A15556-06 A15556-22 A15556-06 A15556-14
AAA15556-06EA 41.69 USD InStock
CAAAA15556-06 AAA15556-06 CAAAA15556-14 CAAAA15556-22 AAA15556-22 AAA15556-14
2-Nitro-4,5-xylidine 97%
2-Nitro-4,5-xylidine


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 138…143 °C
Density: 1.398 g/cm³ (–123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007811
CAS Number: 6972-71-0
EINECS: 230-211-5
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty